Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGTCTCTGCCCATGGGAACTCAC[A/C]ATCTAACAGCGAGACGAGGCACAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609742 MIM: 605815 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL4I1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL4I1 - interleukin 4 induced 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258017.1 | 2852 | Intron | NP_001244946.1 | |||
NM_001258018.1 | 2852 | Intron | NP_001244947.1 | |||
NM_152899.1 | 2852 | Intron | NP_690863.1 | |||
NM_172374.2 | 2852 | Intron | NP_758962.1 |
NUP62 - nucleoporin 62 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193357.1 | 2852 | UTR 3 | NP_001180286.1 | |||
NM_012346.4 | 2852 | UTR 3 | NP_036478.2 | |||
NM_016553.4 | 2852 | UTR 3 | NP_057637.2 | |||
NM_153718.3 | 2852 | UTR 3 | NP_714940.1 | |||
NM_153719.3 | 2852 | UTR 3 | NP_714941.1 |