Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGACTGAAGATAAATACAGTGTCA[A/G]TAGTATCAAGGGACTAGCCCATATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF549 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF549 - zinc finger protein 549 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF550 - zinc finger protein 550 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277090.1 | 4604 | UTR 3 | NP_001264019.1 | |||
NM_001277091.1 | 4604 | UTR 3 | NP_001264020.1 | |||
NM_001277092.1 | 4604 | UTR 3 | NP_001264021.1 | |||
NM_001277093.1 | 4604 | UTR 3 | NP_001264022.1 | |||
XM_011526567.2 | 4604 | UTR 3 | XP_011524869.1 | |||
XM_011526568.2 | 4604 | UTR 3 | XP_011524870.1 | |||
XM_017026401.1 | 4604 | UTR 3 | XP_016881890.1 |