Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGTATTAAATACTAGACAAGGAG[A/C]CAACTCCCGAGGCAGGGTGCTATGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614355 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACSS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACSS1 - acyl-CoA synthetase short-chain family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252675.1 | Intron | NP_001239604.1 | ||||
NM_001252676.1 | Intron | NP_001239605.1 | ||||
NM_001252677.1 | Intron | NP_001239606.1 | ||||
NM_032501.3 | Intron | NP_115890.2 | ||||
XM_006723659.1 | Intron | XP_006723722.1 | ||||
XM_011529388.1 | Intron | XP_011527690.1 |