Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCAGGGTGTAAGATTTTGCGTAAG[A/G]ATTGTTTCCTTTCAAATGGCCCCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602523 | ||||||||||||||||||||
Literature Links: |
DSCAM PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DSCAM - DS cell adhesion molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271534.1 | 6462 | Missense Mutation | CCT,TCT | P,S 1986 | NP_001258463.1 | |
NM_001389.3 | 6462 | Missense Mutation | CCT,TCT | P,S 2004 | NP_001380.2 | |
XM_017028281.1 | 6462 | Missense Mutation | CCT,TCT | P,S 1768 | XP_016883770.1 |