Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTCTCCAGCCCTTGTTGCTGCC[C/T]CCAAGTCCCTTGGCTCTTCCCCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300470 MIM: 300662 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAGED2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAGED2 - MAGE family member D2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014599.5 | Intron | NP_055414.2 | ||||
NM_177433.2 | Intron | NP_803182.1 | ||||
NM_201222.2 | Intron | NP_957516.1 |
SNORA11 - small nucleolar RNA, H/ACA box 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |