Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTAGATCTTTTTACTTCACCTCA[C/T]ACTTATCACCAATGTATATCTCCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300320 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NXT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NXT2 - nuclear transport factor 2 like export factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242617.1 | 1565 | UTR 3 | NP_001229546.1 | |||
NM_001242618.1 | 1565 | UTR 3 | NP_001229547.1 | |||
NM_018698.4 | 1565 | UTR 3 | NP_061168.2 | |||
XM_011530989.2 | 1565 | UTR 3 | XP_011529291.1 | |||
XM_017029648.1 | 1565 | UTR 3 | XP_016885137.1 |