Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCTCTAAAGCTGAATAGGTAACT[A/C]CATATTTCTTTCTTCTGGTTAATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300553 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRWD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BRWD3 - bromodomain and WD repeat domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153252.4 | 10805 | UTR 3 | NP_694984.4 | |||
XM_005262113.3 | 10805 | UTR 3 | XP_005262170.1 | |||
XM_017029384.1 | 10805 | Intron | XP_016884873.1 | |||
XM_017029385.1 | 10805 | Intron | XP_016884874.1 |