Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCATGATCTTAATGGAGGGCAGCC[A/G]CGCCCCGGAGGAGGCAATCTGGGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300892 MIM: 300341 | ||||||||||||||||||||
Literature Links: |
LINC00850 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LINC00850 - long intergenic non-protein coding RNA 850 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAGEA8 - MAGE family member A8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166400.1 | 1174 | Missense Mutation | CAC,CGC | H,R 218 | NP_001159872.1 | |
NM_001166401.1 | 1174 | Missense Mutation | CAC,CGC | H,R 218 | NP_001159873.1 | |
NM_005364.4 | 1174 | Missense Mutation | CAC,CGC | H,R 218 | NP_005355.2 | |
XM_017029520.1 | 1174 | Missense Mutation | CAC,CGC | H,R 218 | XP_016885009.1 |
MAGEA8-AS1 - MAGEA8 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |