Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGTTGCTGCAAGTAACAGCCCCA[A/G]ATACGGAGGGGAAGGCTTCATCGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300380 | ||||||||||||||||||||
Literature Links: |
CTPS2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CTPS2 - CTP synthase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144002.1 | 2365 | Silent Mutation | CTG,TTG | L,L 541 | NP_001137474.1 | |
NM_019857.4 | 2365 | Silent Mutation | CTG,TTG | L,L 541 | NP_062831.3 | |
NM_175859.2 | 2365 | Silent Mutation | CTG,TTG | L,L 541 | NP_787055.1 | |
XM_005274562.3 | 2365 | Silent Mutation | CTG,TTG | L,L 541 | XP_005274619.1 | |
XM_006724503.3 | 2365 | Silent Mutation | CTG,TTG | L,L 541 | XP_006724566.1 | |
XM_011545545.2 | 2365 | Silent Mutation | CTG,TTG | L,L 583 | XP_011543847.1 |