Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATTTTCACATCAAATTGAAACTG[C/T]TATAATTAATTAAATTAAATTAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300753 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APOO PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APOO - apolipoprotein O | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024122.4 | Intron | NP_077027.1 | ||||
XM_011545584.2 | Intron | XP_011543886.1 | ||||
XM_011545585.2 | Intron | XP_011543887.1 | ||||
XM_011545586.2 | Intron | XP_011543888.1 | ||||
XM_011545587.2 | Intron | XP_011543889.1 | ||||
XM_017029837.1 | Intron | XP_016885326.1 |