Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAGGCTAAAGCTGGTGAGAAGTA[G/T]AAATATGAAGTAAGTAACCATTTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 312040 | ||||||||||||||||||||
Literature Links: |
POLA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
POLA1 - DNA polymerase alpha 1, catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016937.3 | 156 | Nonsense Mutation | TAG,TAT | *,Y 47 | NP_058633.2 | |
XM_005274552.3 | 156 | Nonsense Mutation | TAG,TAT | *,Y 53 | XP_005274609.1 | |
XM_006724499.2 | 156 | UTR 5 | XP_006724562.1 | |||
XM_011545540.2 | 156 | Nonsense Mutation | TAG,TAT | *,Y 53 | XP_011543842.1 | |
XM_011545541.2 | 156 | Nonsense Mutation | TAG,TAT | *,Y 53 | XP_011543843.1 | |
XM_017029594.1 | 156 | Nonsense Mutation | TAG,TAT | *,Y 53 | XP_016885083.1 | |
XM_017029595.1 | 156 | Nonsense Mutation | TAG,TAT | *,Y 53 | XP_016885084.1 | |
XM_017029596.1 | 156 | Nonsense Mutation | TAG,TAT | *,Y 53 | XP_016885085.1 |