Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATAACCACAGCAGAATTTTGATAT[A/G]TAATTTAGCCTGTCTTTATATATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300079 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
XIAP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
XIAP - X-linked inhibitor of apoptosis | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167.3 | Intron | NP_001158.2 | ||||
NM_001204401.1 | Intron | NP_001191330.1 | ||||
XM_006724754.2 | Intron | XP_006724817.1 | ||||
XM_011531329.2 | Intron | XP_011529631.1 |