Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAAAGAATACATAAAATAGGAT[A/G]TAATTAAATTCTCATTTATTTAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603756 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCG2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCG2 - ATP binding cassette subfamily G member 2 (Junior blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257386.1 | 4539 | UTR 3 | NP_001244315.1 | |||
NM_004827.2 | 4539 | UTR 3 | NP_004818.2 | |||
XM_005263354.3 | 4539 | UTR 3 | XP_005263411.1 | |||
XM_005263355.3 | 4539 | UTR 3 | XP_005263412.1 | |||
XM_005263356.3 | 4539 | UTR 3 | XP_005263413.1 | |||
XM_011532420.2 | 4539 | UTR 3 | XP_011530722.1 | |||
XM_017008852.1 | 4539 | UTR 3 | XP_016864341.1 | |||
XM_017008853.1 | 4539 | UTR 3 | XP_016864342.1 |