Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGAAGATTCCAGCCACGGGCCCC[A/G]CGTGGAGCACAAACATGCCAGAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608407 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPYSL4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DPYSL4 - dihydropyrimidinase like 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STK32C - serine/threonine kinase 32C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318878.1 | Intron | NP_001305807.1 | ||||
NM_001318879.1 | Intron | NP_001305808.1 | ||||
NM_001318880.1 | Intron | NP_001305809.1 | ||||
NM_001318881.1 | Intron | NP_001305810.1 | ||||
NM_001318882.1 | Intron | NP_001305811.1 | ||||
NM_001318883.1 | Intron | NP_001305812.1 | ||||
NM_173575.3 | Intron | NP_775846.2 | ||||
XM_011539688.1 | Intron | XP_011537990.1 | ||||
XM_011539690.2 | Intron | XP_011537992.1 | ||||
XM_011539693.2 | Intron | XP_011537995.1 | ||||
XM_011539694.1 | Intron | XP_011537996.1 | ||||
XM_011539695.1 | Intron | XP_011537997.1 | ||||
XM_011539696.1 | Intron | XP_011537998.1 | ||||
XM_017016100.1 | Intron | XP_016871589.1 | ||||
XM_017016101.1 | Intron | XP_016871590.1 |