Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAGTCTCCTTTGAGTCATCTTGT[C/T]ACCTTTGGACGTGGATAATGCTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605484 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FCAMR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FCAMR - Fc fragment of IgA and IgM receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122979.2 | 2139 | UTR 3 | NP_001116451.1 | |||
NM_001170631.1 | 2139 | UTR 3 | NP_001164102.1 | |||
NM_032029.4 | 2139 | UTR 3 | NP_114418.2 | |||
XM_011510051.2 | 2139 | UTR 3 | XP_011508353.1 | |||
XM_011510052.2 | 2139 | UTR 3 | XP_011508354.1 | |||
XM_017002442.1 | 2139 | UTR 3 | XP_016857931.1 |