Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGCCGGGCGGGGCCGAGAATTA[C/T]GTAAGACACTGGGCGAGATCTTGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611680 | ||||||||||||||||||||
Literature Links: |
C1orf116 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C1orf116 - chromosome 1 open reading frame 116 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083924.1 | 1500 | Missense Mutation | CAT,CGT | H,R 278 | NP_001077393.1 | |
NM_023938.5 | 1500 | Missense Mutation | CAT,CGT | H,R 524 | NP_076427.2 | |
XM_005273259.1 | 1500 | Missense Mutation | CAT,CGT | H,R 278 | XP_005273316.1 | |
XM_006711530.1 | 1500 | Missense Mutation | CAT,CGT | H,R 524 | XP_006711593.1 | |
XM_011509973.2 | 1500 | Missense Mutation | CAT,CGT | H,R 278 | XP_011508275.1 |