Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGATCAGGCACATGGTAAGGGTGA[C/T]GTGTGGCATTTGTTTTGTGTAATCA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 601470 | |||||||||||||||||||||||
Literature Links: |
CX3CR1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CX3CR1 - C-X3-C motif chemokine receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171171.1 | 1846 | UTR 3 | NP_001164642.1 | |||
NM_001171172.1 | 1846 | UTR 3 | NP_001164643.1 | |||
NM_001171174.1 | 1846 | UTR 3 | NP_001164645.1 | |||
NM_001337.3 | 1846 | UTR 3 | NP_001328.1 |