Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCCCATGAAGCTGAAGTTGGGAG[A/C]GATGATGCTTCGCCTCTGCTTCACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600714 | ||||||||||||||||||||
Literature Links: |
DUSP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DUSP1 - dual specificity phosphatase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004417.3 | 1134 | Missense Mutation | GCT,TCT | A,S 296 | NP_004408.1 |
LOC101928093 - uncharacterized LOC101928093 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |