Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGTGTTACCTTCAACCACTTGTC[A/T]ACACACTTGGTGTGGAACTCATGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
RNF44 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RNF44 - ring finger protein 44 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014901.4 | 1637 | Silent Mutation | GTA,GTT | V,V 407 | NP_055716.1 | |
XM_005265840.2 | 1637 | Silent Mutation | GTA,GTT | V,V 407 | XP_005265897.1 | |
XM_005265841.4 | 1637 | Silent Mutation | GTA,GTT | V,V 407 | XP_005265898.1 | |
XM_005265842.4 | 1637 | Silent Mutation | GTA,GTT | V,V 326 | XP_005265899.1 | |
XM_005265843.2 | 1637 | Silent Mutation | GTA,GTT | V,V 326 | XP_005265900.1 | |
XM_005265844.3 | 1637 | Silent Mutation | GTA,GTT | V,V 326 | XP_005265901.1 | |
XM_005265845.4 | 1637 | Silent Mutation | GTA,GTT | V,V 326 | XP_005265902.1 | |
XM_006714831.2 | 1637 | Silent Mutation | GTA,GTT | V,V 404 | XP_006714894.1 | |
XM_006714832.2 | 1637 | Silent Mutation | GTA,GTT | V,V 360 | XP_006714895.1 | |
XM_011534466.2 | 1637 | Silent Mutation | GTA,GTT | V,V 407 | XP_011532768.1 | |
XM_011534467.2 | 1637 | Silent Mutation | GTA,GTT | V,V 326 | XP_011532769.1 |