Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGCAGTACAGTTTTTTTTCTCCA[A/G]AAGACAAAAGTCAGTTTCATCCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610703 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSPH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HSPH1 - heat shock protein family H (Hsp110) member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286503.1 | 2844 | UTR 3 | NP_001273432.1 | |||
NM_001286504.1 | 2844 | UTR 3 | NP_001273433.1 | |||
NM_001286505.1 | 2844 | UTR 3 | NP_001273434.1 | |||
NM_006644.3 | 2844 | UTR 3 | NP_006635.2 | |||
XM_005266236.1 | 2844 | UTR 3 | XP_005266293.1 | |||
XM_011534887.2 | 2844 | UTR 3 | XP_011533189.1 | |||
XM_011534888.1 | 2844 | UTR 3 | XP_011533190.1 | |||
XM_017020361.1 | 2844 | UTR 3 | XP_016875850.1 | |||
XM_017020362.1 | 2844 | UTR 3 | XP_016875851.1 | |||
XM_017020363.1 | 2844 | UTR 3 | XP_016875852.1 | |||
XM_017020364.1 | 2844 | UTR 3 | XP_016875853.1 |