Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCATCAAGTGACTCCCTCTTTTTC[C/T]ACCCTACCAACATCACTCTAATTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607441 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPSTI1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EPSTI1 - epithelial stromal interaction 1 (breast) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005266596.1 | 1749 | UTR 3 | XP_005266653.1 | |||
XM_005266597.1 | 1749 | UTR 3 | XP_005266654.1 | |||
XM_006719896.3 | 1749 | Intron | XP_006719959.1 | |||
XM_011535312.1 | 1749 | UTR 3 | XP_011533614.1 | |||
XM_011535313.1 | 1749 | UTR 3 | XP_011533615.1 | |||
XM_011535314.1 | 1749 | UTR 3 | XP_011533616.1 | |||
XM_017020853.1 | 1749 | UTR 3 | XP_016876342.1 | |||
XM_017020854.1 | 1749 | UTR 3 | XP_016876343.1 | |||
XM_017020855.1 | 1749 | UTR 3 | XP_016876344.1 |