Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCCCGTACCGCAGCCCCCGGGCA[C/T]GACCTCTCGGACACGGTGCAGGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610368 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRCH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRCH1 - leucine rich repeats and calponin homology domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164211.1 | 393 | Silent Mutation | CAC,CAT | H,H 94 | NP_001157683.1 | |
NM_001164213.1 | 393 | Silent Mutation | CAC,CAT | H,H 94 | NP_001157685.1 | |
NM_015116.2 | 393 | Silent Mutation | CAC,CAT | H,H 94 | NP_055931.1 | |
XM_017020483.1 | 393 | Silent Mutation | CAC,CAT | H,H 94 | XP_016875972.1 | |
XM_017020484.1 | 393 | UTR 5 | XP_016875973.1 |