Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCCTTTTTGCAATAGGTTCTTC[C/T]TGCTTTGCTGGTGATCATGTCACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602182 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENPP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ENPP3 - ectonucleotide pyrophosphatase/phosphodiesterase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005021.4 | 236 | Missense Mutation | CCT,CTT | P,L 29 | NP_005012.2 | |
XM_011535897.1 | 236 | Intron | XP_011534199.1 | |||
XM_017010932.1 | 236 | Intron | XP_016866421.1 |