Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGACAGAGGCTGTGTCTACCTGA[A/G]GAACTGGAGAGGTAGAATTAGAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614555 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FRMD6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FRMD6 - FERM domain containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042481.2 | Intron | NP_001035946.1 | ||||
NM_001267046.1 | Intron | NP_001253975.1 | ||||
NM_001267047.1 | Intron | NP_001253976.1 | ||||
NM_152330.3 | Intron | NP_689543.1 | ||||
XM_006720029.1 | Intron | XP_006720092.1 | ||||
XM_006720030.1 | Intron | XP_006720093.1 | ||||
XM_011536423.1 | Intron | XP_011534725.1 | ||||
XM_011536424.1 | Intron | XP_011534726.1 | ||||
XM_017020967.1 | Intron | XP_016876456.1 | ||||
XM_017020968.1 | Intron | XP_016876457.1 |
FRMD6-AS1 - FRMD6 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |