Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGCCGGGACCTGGGGGCTCCTGC[C/T]GGATCCATGGGGGCGGCCAGCTGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602699 | ||||||||||||||||||||
Literature Links: |
NFATC4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NFATC4 - nuclear factor of activated T-cells 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136022.2 | 315 | Silent Mutation | GCC,GCT | A,A 61 | NP_001129494.1 | |
NM_001198965.1 | 315 | UTR 5 | NP_001185894.1 | |||
NM_001198966.2 | 315 | Intron | NP_001185895.1 | |||
NM_001198967.2 | 315 | Silent Mutation | GCC,GCT | A,A 61 | NP_001185896.1 | |
NM_001288802.1 | 315 | Intron | NP_001275731.1 | |||
NM_001320043.1 | 315 | Silent Mutation | GCC,GCT | A,A 61 | NP_001306972.1 | |
NM_004554.4 | 315 | UTR 5 | NP_004545.2 | |||
XM_011536797.2 | 315 | Intron | XP_011535099.1 | |||
XM_011536798.1 | 315 | Intron | XP_011535100.1 | |||
XM_011536799.2 | 315 | Intron | XP_011535101.1 |