Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGTCTATCATCAGCATCCAAAAA[C/T]TATGATTAAAAATTTAGTCAATTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFCAB11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EFCAB11 - EF-hand calcium binding domain 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284266.1 | 693 | Intron | NP_001271195.1 | |||
NM_001284267.1 | 693 | UTR 3 | NP_001271196.1 | |||
NM_001284268.1 | 693 | Intron | NP_001271197.1 | |||
NM_001284269.1 | 693 | UTR 3 | NP_001271198.1 | |||
NM_145231.3 | 693 | UTR 3 | NP_660274.1 |