Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACCCTTGGTCCAGGCTCCTTAGT[C/T]TTGAGAATTCTTACACCGGGTGTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612342 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GGT7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GGT7 - gamma-glutamyltransferase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178026.2 | 1883 | Intron | NP_821158.2 | |||
XM_011528782.2 | 1883 | Intron | XP_011527084.1 | |||
XM_011528783.2 | 1883 | Intron | XP_011527085.1 | |||
XM_017027809.1 | 1883 | Intron | XP_016883298.1 | |||
XM_017027810.1 | 1883 | UTR 3 | XP_016883299.1 | |||
XM_017027811.1 | 1883 | Intron | XP_016883300.1 |