Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAGGTCTTCTGGCCTGGGCTGGG[A/G]GCAGTCACTCAGGGGGCATTTGACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120140 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL2A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COL2A1 - collagen type II alpha 1 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001844.4 | 6325 | UTR 3 | NP_001835.3 | |||
NM_033150.2 | 6325 | UTR 3 | NP_149162.2 | |||
XM_017018828.1 | 6325 | UTR 3 | XP_016874317.1 | |||
XM_017018829.1 | 6325 | UTR 3 | XP_016874318.1 | |||
XM_017018830.1 | 6325 | UTR 3 | XP_016874319.1 | |||
XM_017018831.1 | 6325 | UTR 3 | XP_016874320.1 |
TMEM106C - transmembrane protein 106C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |