Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCACCACGCTCGGGACCCAGTAC[A/G]TTGGGGAAAGGACAGCAGAAGGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612314 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GSTO2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GSTO2 - glutathione S-transferase omega 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ITPRIP - inositol 1,4,5-trisphosphate receptor interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001272012.1 | 2528 | UTR 3 | NP_001258941.1 | |||
NM_001272013.1 | 2528 | UTR 3 | NP_001258942.1 | |||
NM_033397.3 | 2528 | UTR 3 | NP_203755.1 | |||
XM_005270257.2 | 2528 | UTR 3 | XP_005270314.1 |
LOC101927472 - uncharacterized LOC101927472 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |