Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAACCATGACAGTAGGAAGGATGA[C/T]TTGTCTTGCATCAATAGAGCACACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608599 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAB11FIP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAB11FIP2 - RAB11 family interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014904.2 | 2608 | UTR 3 | NP_055719.1 | |||
XM_005269629.3 | 2608 | UTR 3 | XP_005269686.1 | |||
XM_017015926.1 | 2608 | Intron | XP_016871415.1 |