Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTCCTGAGATGACAGATTCCTTT[C/G]TGGCACTGTGTAGCCAACTGGAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123860 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTPS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTPS1 - CTP synthase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLFNL1 - schlafen like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168247.2 | 1892 | UTR 3 | NP_001161719.1 | |||
NM_001300859.1 | 1892 | UTR 3 | NP_001287788.1 | |||
NM_144990.3 | 1892 | UTR 3 | NP_659427.3 | |||
XM_005270597.2 | 1892 | UTR 3 | XP_005270654.1 | |||
XM_005270598.2 | 1892 | UTR 3 | XP_005270655.1 | |||
XM_005270599.2 | 1892 | UTR 3 | XP_005270656.1 | |||
XM_006710432.3 | 1892 | UTR 3 | XP_006710495.1 | |||
XM_011540943.1 | 1892 | UTR 3 | XP_011539245.1 | |||
XM_011540945.1 | 1892 | UTR 3 | XP_011539247.1 | |||
XM_011540947.1 | 1892 | UTR 3 | XP_011539249.1 | |||
XM_011540948.1 | 1892 | UTR 3 | XP_011539250.1 | |||
XM_011540949.1 | 1892 | UTR 3 | XP_011539251.1 | |||
XM_011540950.1 | 1892 | UTR 3 | XP_011539252.1 | |||
XM_011540951.1 | 1892 | UTR 3 | XP_011539253.1 | |||
XM_011540952.2 | 1892 | UTR 3 | XP_011539254.1 | |||
XM_011540953.2 | 1892 | UTR 3 | XP_011539255.1 | |||
XM_011540954.1 | 1892 | UTR 3 | XP_011539256.1 | |||
XM_017000573.1 | 1892 | UTR 3 | XP_016856062.1 | |||
XM_017000574.1 | 1892 | UTR 3 | XP_016856063.1 |
SLFNL1-AS1 - SLFNL1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |