Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGGAGCTGCTGAAGCGTGTGGAC[C/T]TCGCGACGGTCCCGCGGAGACATCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
38 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615394 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NSUN4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NSUN4 - NOP2/Sun RNA methyltransferase family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256127.1 | 702 | Intron | NP_001243056.1 | |||
NM_001256128.1 | 702 | Intron | NP_001243057.1 | |||
NM_199044.3 | 702 | Missense Mutation | CTC,TTC | L,F 18 | NP_950245.2 |