Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
30 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610162 MIM: 608676 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC28B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC28B - coiled-coil domain containing 28B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301011.1 | 1434 | Silent Mutation | TTC,TTT | F,F 110 | NP_001287940.1 | |
NM_024296.4 | 1434 | Silent Mutation | TTC,TTT | F,F 110 | NP_077272.2 | |
XM_011542113.2 | 1434 | Silent Mutation | TTC,TTT | F,F 110 | XP_011540415.1 | |
XM_017002306.1 | 1434 | Silent Mutation | TTC,TTT | F,F 110 | XP_016857795.1 | |
XM_017002307.1 | 1434 | Silent Mutation | TTC,TTT | F,F 110 | XP_016857796.1 | |
XM_017002308.1 | 1434 | Silent Mutation | TTC,TTT | F,F 110 | XP_016857797.1 | |
XM_017002309.1 | 1434 | Silent Mutation | TTC,TTT | F,F 110 | XP_016857798.1 |
DCDC2B - doublecortin domain containing 2B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IQCC - IQ motif containing C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TXLNA - taxilin alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |