Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAGTTGCGGAAAGATATCCGACC[C/T]GAATATCGAGAGGATTTTGTTCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600727 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NFIA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NFIA - nuclear factor I A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134673.3 | 661 | Silent Mutation | CCC,CCT | P,P 85 | NP_001128145.1 | |
NM_001145511.1 | 661 | Silent Mutation | CCC,CCT | P,P 77 | NP_001138983.1 | |
NM_001145512.1 | 661 | Silent Mutation | CCC,CCT | P,P 130 | NP_001138984.1 | |
NM_005595.4 | 661 | Silent Mutation | CCC,CCT | P,P 85 | NP_005586.1 | |
XM_011541512.2 | 661 | Silent Mutation | CCC,CCT | P,P 85 | XP_011539814.1 | |
XM_011541514.2 | 661 | Silent Mutation | CCC,CCT | P,P 77 | XP_011539816.1 | |
XM_011541515.2 | 661 | Silent Mutation | CCC,CCT | P,P 85 | XP_011539817.1 | |
XM_017001362.1 | 661 | Silent Mutation | CCC,CCT | P,P 77 | XP_016856851.1 | |
XM_017001363.1 | 661 | Intron | XP_016856852.1 |