Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCACAGCTTCTCTGATGTTCCTT[A/G]GTCTGAAAGACACAGCTTGAGTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
24 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611906 MIM: 611889 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FNDC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FNDC5 - fibronectin type III domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171940.1 | 1619 | UTR 3 | NP_001165411.2 | |||
NM_001171941.2 | 1619 | UTR 3 | NP_001165412.1 | |||
NM_153756.2 | 1619 | UTR 3 | NP_715637.2 |
S100PBP - S100P binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |