Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCAAGAAGTTACAATATAATGG[A/G]AAAATAAATCGGCCCATACATTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615684 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HFM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HFM1 - HFM1, ATP dependent DNA helicase homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017975.4 | 3622 | UTR 3 | NP_001017975.4 | |||
XM_011540849.1 | 3622 | UTR 3 | XP_011539151.1 | |||
XM_011540850.2 | 3622 | UTR 3 | XP_011539152.1 | |||
XM_011540851.1 | 3622 | UTR 3 | XP_011539153.1 | |||
XM_011540852.2 | 3622 | UTR 3 | XP_011539154.1 | |||
XM_011540855.1 | 3622 | UTR 3 | XP_011539157.1 | |||
XM_011540857.1 | 3622 | UTR 3 | XP_011539159.1 | |||
XM_011540859.2 | 3622 | UTR 3 | XP_011539161.1 | |||
XM_017000490.1 | 3622 | UTR 3 | XP_016855979.1 | |||
XM_017000491.1 | 3622 | UTR 3 | XP_016855980.1 | |||
XM_017000492.1 | 3622 | UTR 3 | XP_016855981.1 | |||
XM_017000493.1 | 3622 | UTR 3 | XP_016855982.1 | |||
XM_017000494.1 | 3622 | Intron | XP_016855983.1 | |||
XM_017000495.1 | 3622 | Intron | XP_016855984.1 | |||
XM_017000496.1 | 3622 | Intron | XP_016855985.1 |