Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAAACAATAATTTCAAAGCTTCT[G/A]GTCTCCTTTCAGATTTACTTTGGTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
30 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611199 MIM: 601176 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNTTIP2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DNTTIP2 - deoxynucleotidyltransferase terminal interacting protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GCLM - glutamate-cysteine ligase modifier subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308253.1 | 1864 | UTR 3 | NP_001295182.1 | |||
NM_002061.3 | 1864 | UTR 3 | NP_002052.1 | |||
XM_011541261.2 | 1864 | UTR 3 | XP_011539563.1 | |||
XM_017001056.1 | 1864 | UTR 3 | XP_016856545.1 | |||
XM_017001057.1 | 1864 | UTR 3 | XP_016856546.1 |
Set Membership: |
HapMap |