Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGATACAGTTCATACAATCTAAA[A/T]AGATACAGTCTTTACTTTTATTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602665 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CASP5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CASP5 - caspase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136109.1 | Intron | NP_001129581.1 | ||||
NM_001136110.1 | Intron | NP_001129582.1 | ||||
NM_001136112.1 | Intron | NP_001129584.1 | ||||
NM_004347.3 | Intron | NP_004338.3 | ||||
XM_011543021.2 | Intron | XP_011541323.1 |