Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTCTTTGACCGAGAGATTGTGGT[C/T]ACCATCAGCTGGGCCAAGAGCATCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601998 MIM: 606583 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ESRRA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ESRRA - estrogen related receptor alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282450.1 | 932 | Intron | NP_001269379.1 | |||
NM_001282451.1 | 932 | Silent Mutation | GTC,GTT | V,V 237 | NP_001269380.1 | |
NM_004451.4 | 932 | Silent Mutation | GTC,GTT | V,V 238 | NP_004442.3 | |
XM_017017313.1 | 932 | Silent Mutation | GTC,GTT | V,V 248 | XP_016872802.1 |
PRDX5 - peroxiredoxin 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRMT112 - tRNA methyltransferase 11-2 homolog (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |