Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTCCCAGGTGGCAAATGCAAAT[A/G]CAAGAGCAGCCAGAAGCAGCACAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616926 MIM: 601037 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FXYD4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FXYD4 - FXYD domain containing ion transport regulator 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184963.1 | 520 | Missense Mutation | TAC,TGC | Y,C 63 | NP_001171892.1 | |
NM_173160.2 | 520 | Missense Mutation | TAC,TGC | Y,C 63 | NP_775183.1 |
HNRNPF - heterogeneous nuclear ribonucleoprotein F | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |