Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGCTACTTAGCTGGGGTCTCTTA[C/T]CAAGATAGTAAGAAAACGGCATGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614620 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRAMP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
CRAMP1 - cramped chromatin regulator homolog 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020825.3 | Intron | NP_065876.3 |
IFT140 - intraflagellar transport 140 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105371046 - uncharacterized LOC105371046 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |