Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTGGCCCTGCGGGAGCACCCTGC[C/G]CTGCTGGTGGAATTCTGTGAGTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602844 MIM: 603816 MIM: 608012 MIM: 603895 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGDIG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGDIG - Rho GDP dissociation inhibitor gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001176.3 | 235 | Intron | NP_001167.2 |
AXIN1 - axin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDIA2 - protein disulfide isomerase family A member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006849.2 | 235 | Silent Mutation | GCC,GCG | A,A 61 | NP_006840.2 |
RGS11 - regulator of G-protein signaling 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |