Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTTTGCTACTTAACCTAATTGT[A/G]TTACTTTGTTCTCTTGCAGTTCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
29 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613562 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FCRL6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FCRL6 - Fc receptor like 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004310.2 | Intron | NP_001004310.2 | ||||
NM_001284217.1 | Intron | NP_001271146.1 | ||||
XM_005245128.3 | Intron | XP_005245185.1 | ||||
XM_005245129.3 | Intron | XP_005245186.1 | ||||
XM_005245131.3 | Intron | XP_005245188.1 | ||||
XM_006711292.3 | Intron | XP_006711355.1 | ||||
XM_011509480.1 | Intron | XP_011507782.1 | ||||
XM_011509481.2 | Intron | XP_011507783.1 | ||||
XM_017001176.1 | Intron | XP_016856665.1 | ||||
XM_017001177.1 | Intron | XP_016856666.1 |