Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCGATTGTCACACGGCCGTTCTC[C/T]CTGCCAAATCCTGAAAGGAGGAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602733 MIM: 604564 | ||||||||||||||||||||
Literature Links: |
ALDH9A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ALDH9A1 - aldehyde dehydrogenase 9 family member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000696.3 | 1578 | Silent Mutation | AGA,AGG | R,R 491 | NP_000687.3 | |
XM_011509294.2 | 1578 | Silent Mutation | AGA,AGG | R,R 397 | XP_011507596.1 |
MGST3 - microsomal glutathione S-transferase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |