Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGGTAGGCTTACCTCCAAATAC[A/G]AAAAAGCTGAGAGGCTCCTGCTGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 605205 MIM: 614737 | ||||||||||||||||||||
Literature Links: |
ADCY10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADCY10 - adenylate cyclase 10, soluble | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MPC2 - mitochondrial pyruvate carrier 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143674.3 | 525 | Missense Mutation | CGT,TGT | R,C 113 | NP_001137146.1 | |
NM_015415.3 | 525 | Missense Mutation | CGT,TGT | R,C 113 | NP_056230.1 | |
XM_006711266.3 | 525 | Missense Mutation | CGT,TGT | R,C 113 | XP_006711329.1 |