Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGTAGGGAGTGAAGTCTCAGATA[A/G]GAGGACCTGTGTGAGCCTCACTACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
17 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600250 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
XCL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
XCL1 - X-C motif chemokine ligand 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002995.2 | 251 | Missense Mutation | AAG,AGG | K,R 29 | NP_002986.1 | |
XM_011509865.2 | 251 | Intron | XP_011508167.1 |