Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAATATGTTGCCTAAAATCTGTAG[A/G]GGCCCATTGGGCATTCTGCTTCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
29 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MROH9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MROH9 - maestro heat like repeat family member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163629.1 | Intron | NP_001157101.1 | ||||
NM_025063.2 | Intron | NP_079339.2 | ||||
XM_011510005.2 | Intron | XP_011508307.1 | ||||
XM_011510006.2 | Intron | XP_011508308.1 | ||||
XM_011510007.2 | Intron | XP_011508309.1 |