Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAGGCCACCACCACCACCGCCAC[C/T]GCCACCACTACCACCTCCGCCGCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 134638 | ||||||||||||||||||||
Literature Links: |
FASLG PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FASLG - Fas ligand | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000639.2 | 346 | Missense Mutation | CCG,CTG | P,L 51 | NP_000630.1 | |
NM_001302746.1 | 346 | Missense Mutation | CCG,CTG | P,L 51 | NP_001289675.1 |