Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTTTCTGCTTGTTGCTGTGTGC[A/G]GGTGTTCGGCCGCGATCACCTTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601013 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CACNA1E PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CACNA1E - calcium voltage-gated channel subunit alpha1 E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000721.3 | 143 | UTR 5 | NP_000712.2 | |||
NM_001205293.1 | 143 | UTR 5 | NP_001192222.1 | |||
NM_001205294.1 | 143 | UTR 5 | NP_001192223.1 | |||
XM_017002243.1 | 143 | Intron | XP_016857732.1 | |||
XM_017002244.1 | 143 | Intron | XP_016857733.1 | |||
XM_017002245.1 | 143 | Intron | XP_016857734.1 | |||
XM_017002246.1 | 143 | Intron | XP_016857735.1 | |||
XM_017002247.1 | 143 | Intron | XP_016857736.1 | |||
XM_017002248.1 | 143 | Intron | XP_016857737.1 | |||
XM_017002249.1 | 143 | Intron | XP_016857738.1 | |||
XM_017002250.1 | 143 | Intron | XP_016857739.1 | |||
XM_017002251.1 | 143 | Intron | XP_016857740.1 |