Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAGATGCACAAAGCAATGCTGC[A/C]AGAGACTTTGTTAACTATTTGGTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 603115 | ||||||||||||||||||||
Literature Links: |
DHX9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DHX9 - DEAH-box helicase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001357.4 | 361 | Silent Mutation | GCA,GCC | A,A 62 | NP_001348.2 |
LOC647070 - nascent polypeptide-associated complex subunit alpha, muscle-specific form-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |